Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrm1
Entrez Gene ID
363661
Gene Name
mitochondrial rRNA methyltransferase 1 homolog (S. cerevisiae)
Gene Aliases
RGD1566232
Chromosome Location
Chr.10: 72136246 - 72142533 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108832.1 NP_001102302.1 3 - 4 905 97

View Full Product Details