Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Skp1
Entrez Gene ID
287280
Gene Name
S-phase kinase-associated protein 1
Gene Aliases
Skp1a
Chromosome Location
Chr.10: 37594578 - 37609653 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007608.2 NP_001007609.1 1 - 2 80 103
GenBank mRNA BC058152.1 - 1 - 2 83 103

View Full Product Details