Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klf12
Entrez Gene ID
306110
Gene Name
Kruppel-like factor 12
Gene Aliases
-
Chromosome Location
Chr.15: 84316986 - 84748539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107281.1 NP_001100751.1 2 - 3 203 143
XM_006252408.2 XP_006252470.1 2 - 3 205 143

View Full Product Details