Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klf12
Entrez Gene ID
306110
Gene Name
Kruppel-like factor 12
Gene Aliases
-
Chromosome Location
Chr.15: 84316986 - 84748539 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006252408.2 XP_006252470.1 4 - 5 839 129
XM_006252409.2 XP_006252471.1 4 - 5 834 129
XM_006252410.2 XP_006252472.1 4 - 5 905 129
XM_006252413.2 XP_006252475.1 3 - 4 687 129
XM_006252415.2 XP_006252477.1 3 - 4 684 129
XM_008770870.1 XP_008769092.1 3 - 4 2641 129
XM_008770871.1 XP_008769093.1 5 - 6 851 129

View Full Product Details