Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpc6
Entrez Gene ID
691984
Gene Name
glypican 6
Gene Aliases
-
Chromosome Location
Chr.15: 102164183 - 103174721 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001077762.5 XP_001077762.3 7 - 8 2167 69
XM_003751533.3 XP_003751581.1 8 - 9 2198 69

View Full Product Details