The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017342.2 | NP_059038.1 | 4 - 5 | 450 | 129 |
GenBank mRNA | BC072693.1 | - | 4 - 5 | 443 | 129 |
X14221.1 | - | 4 - 5 | 451 | 129 |