Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a11
Entrez Gene ID
287796
Gene Name
solute carrier family 39, member 11
Gene Aliases
-
Chromosome Location
Chr.10: 101754842 - 102169099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013042.1 NP_001013060.1 5 - 6 540 77
XM_006247665.2 XP_006247727.1 5 - 6 838 77
GenBank mRNA BC062054.1 - 5 - 6 540 77

View Full Product Details