Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc39a11
Entrez Gene ID
287796
Gene Name
solute carrier family 39, member 11
Gene Aliases
-
Chromosome Location
Chr.10: 101754842 - 102169099 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247670.2 XP_006247732.1 1 - 2 944 112

View Full Product Details