Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Luc7l
Entrez Gene ID
360503
Gene Name
LUC7-like (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.10: 15538880 - 15572651 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024269.1 NP_001019440.1 6 - 7 943 64
XM_006246006.2 XP_006246068.1 7 - 8 908 64
XM_006246007.1 XP_006246069.1 7 - 8 2312 64
XM_008767588.1 XP_008765810.1 7 - 8 1771 64
GenBank mRNA BC089960.1 - 6 - 7 943 64

View Full Product Details