The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024355.1 | NP_077331.1 | 9 - 10 | 2474 | 94 |
GenBank mRNA | AF017757.1 | - | 9 - 10 | 2474 | 94 |