Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a4
Entrez Gene ID
64037
Gene Name
solute carrier family 22 (organic cation/zwitterion transporter), member 4
Gene Aliases
Octn1
Chromosome Location
Chr.10: 39334972 - 39373508 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022270.1 NP_071606.1 - 1127 104
GenBank mRNA AF169831.1 - - 1127 104

View Full Product Details