The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008818.1 | NP_001008818.1 | 5 - 6 | 1011 | 108 |
XM_006247394.2 | XP_006247456.1 | 5 - 6 | 1035 | 108 |