The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014120.1 | NP_001014142.1 | 1 - 2 | 292 | 74 |
GenBank mRNA | BC085854.1 | - | 1 - 2 | 292 | 74 |