Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nkx2-8
Entrez Gene ID
299061
Gene Name
NK2 homeobox 8
Gene Aliases
Nkx2-9
Chromosome Location
Chr.6: 77506882 - 77508585 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106732.1 NP_001100202.1 1 - 2 370 99

View Full Product Details