The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001100492.1 | NP_001093962.1 | 2 - 2 | 263 | 101 |
NM_001185047.1 | NP_001171976.1 | 3 - 3 | 320 | 101 | |
GenBank mRNA | AY245001.1 | - | 2 - 2 | 201 | 101 |
CB698538.1 | - | - | 330 | 101 | |
FM050402.1 | - | - | 263 | 101 |