Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rnf43
Entrez Gene ID
303412
Gene Name
ring finger protein 43
Gene Aliases
RGD1305204
Chromosome Location
Chr.10: 74956889 - 75028951 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135921.1 NP_001129393.1 4 - 5 588 72
XM_006247090.2 XP_006247152.1 5 - 6 2073 72
XM_006247091.2 XP_006247153.1 5 - 6 1848 72
XM_006247092.2 XP_006247154.1 4 - 5 2593 72
XM_006247094.2 XP_006247156.1 3 - 4 1949 72
XM_008768038.1 XP_008766260.1 4 - 5 2984 72
XM_008768039.1 XP_008766261.1 2 - 3 2627 72
XM_008768040.1 XP_008766262.1 3 - 4 2529 72
XM_008768041.1 XP_008766263.1 3 - 4 2177 72

View Full Product Details