The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012751.1 | NP_036883.1 | 10 - 11 | 1468 | 68 |
XM_006246596.2 | XP_006246658.1 | 11 - 12 | 1441 | 68 | |
GenBank mRNA | BC085757.1 | - | 10 - 11 | 1513 | 68 |
D28561.1 | - | 10 - 11 | 1468 | 68 | |
J04524.1 | - | 10 - 11 | 1450 | 68 | |
M25482.1 | - | 10 - 11 | 1433 | 68 | |
X14771.1 | - | 10 - 11 | 1481 | 68 |