Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a4
Entrez Gene ID
25139
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 4
Gene Aliases
Glut4
Chromosome Location
Chr.10: 56552983 - 56558561 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012751.1 NP_036883.1 8 - 9 1158 132
XM_006246596.2 XP_006246658.1 9 - 10 1131 132
GenBank mRNA BC085757.1 - 8 - 9 1203 132
D28561.1 - 8 - 9 1158 132
J04524.1 - 8 - 9 1140 132
M25482.1 - 8 - 9 1123 132
X14771.1 - 8 - 9 1171 132

View Full Product Details