The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014274.1 | NP_001014296.1 | 4 - 5 | 461 | 97 |
XM_008773429.1 | XP_008771651.1 | 4 - 5 | 571 | 97 | |
GenBank mRNA | BC081886.1 | - | 4 - 5 | 461 | 97 |