Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plin2
Entrez Gene ID
298199
Gene Name
perilipin 2
Gene Aliases
Adfp, Adrp
Chromosome Location
Chr.5: 104984413 - 105010853 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007144.1 NP_001007145.1 4 - 5 393 120
XM_006238360.2 XP_006238422.1 4 - 5 429 120
XM_006238362.2 XP_006238424.1 3 - 4 251 120
XM_008763778.1 XP_008762000.1 4 - 5 429 120
GenBank mRNA BC085861.1 - 4 - 5 393 120

View Full Product Details