Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Khk
Entrez Gene ID
25659
Gene Name
ketohexokinase
Gene Aliases
KETHPRO
Chromosome Location
Chr.6: 26810577 - 26821011 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031855.3 NP_114061.1 6 - 7 855 114
XM_006239790.2 XP_006239852.1 6 - 7 1055 114
XM_008764507.1 XP_008762729.1 6 - 7 1074 114
GenBank mRNA BC078752.1 - 6 - 7 855 114
X63658.1 - 6 - 7 915 114

View Full Product Details