Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mycn
Entrez Gene ID
298894
Gene Name
v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
Gene Aliases
N-myc, Nmuc1, Nmyc
Chromosome Location
Chr.6: 38222554 - 38228406 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013096.1 NP_001013114.1 2 - 3 1075 95

View Full Product Details