Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem18
Entrez Gene ID
362722
Gene Name
transmembrane protein 18
Gene Aliases
-
Chromosome Location
Chr.6: 49529059 - 49536677 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007748.1 NP_001007749.1 3 - 4 343 81
XM_008764598.1 XP_008762820.1 1 - 2 753 81
XM_008764599.1 XP_008762821.1 2 - 3 618 81
GenBank mRNA BC076388.1 - 3 - 4 343 81

View Full Product Details