The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105720.2 | NP_001099190.2 | 2 - 3 | 455 | 73 |
GenBank mRNA | BC166886.1 | - | 2 - 3 | 407 | 73 |
CB316505.1 | - | 2 - 3 | 409 | 73 | |
FQ215246.1 | - | 2 - 3 | 407 | 73 | |
FQ225350.1 | - | 2 - 3 | 402 | 73 | |
FQ225877.1 | - | 2 - 3 | 405 | 73 | |
FQ226288.1 | - | 2 - 3 | 405 | 73 | |
FQ227283.1 | - | 2 - 3 | 405 | 73 | |
FQ233200.1 | - | 2 - 3 | 405 | 73 | |
FQ233627.1 | - | 2 - 3 | 393 | 73 | |
FQ234262.1 | - | 2 - 3 | 407 | 73 | |
X63594.1 | - | 2 - 3 | 385 | 73 |