Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lpar4
Entrez Gene ID
302378
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
Gpr23, RGD1563686
Chromosome Location
Chr.X: 78042859 - 78056347 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106940.1 NP_001100410.1 1 - 2 377 75

View Full Product Details