The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017242.1 | NP_058938.1 | 4 - 5 | 710 | 72 |
XM_006248339.2 | XP_006248401.1 | 4 - 5 | 1161 | 72 | |
GenBank mRNA | AY326256.1 | - | 4 - 5 | 679 | 72 |
BC087607.1 | - | 4 - 5 | 681 | 72 | |
U31554.1 | - | 4 - 5 | 710 | 72 |