Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lsamp
Entrez Gene ID
29561
Gene Name
limbic system-associated membrane protein
Gene Aliases
-
Chromosome Location
Chr.11: 58795888 - 59442772 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248339.2 XP_006248401.1 8 - 9 1497 88
GenBank mRNA AY326256.1 - 8 - 9 1015 88
BC087607.1 - 8 - 9 1017 88

View Full Product Details