Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hcls1
Entrez Gene ID
288077
Gene Name
hematopoietic cell specific Lyn substrate 1
Gene Aliases
-
Chromosome Location
Chr.11: 66736966 - 66759402 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011898.1 NP_001011898.1 1 - 2 33 64
GenBank mRNA BC079081.1 - 1 - 2 33 64

View Full Product Details