Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem39a
Entrez Gene ID
288092
Gene Name
transmembrane protein 39a
Gene Aliases
RGD1306421
Chromosome Location
Chr.11: 64723128 - 64752553 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013865.1 NP_001013887.1 2 - 3 468 115
XM_008768717.1 XP_008766939.1 2 - 3 507 115
GenBank mRNA BC085845.1 - 2 - 3 468 115

View Full Product Details