Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sash3
Entrez Gene ID
317578
Gene Name
SAM and SH3 domain containing 3
Gene Aliases
RGD1560293
Chromosome Location
Chr.X: 134979636 - 134994351 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134992.1 NP_001128464.1 5 - 6 742 70
XM_006257530.2 XP_006257592.1 5 - 6 796 70
GenBank mRNA BC169091.1 - 5 - 6 742 70
FQ226243.1 - 1 - 2 44 70

View Full Product Details