Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Frmd7
Entrez Gene ID
302826
Gene Name
FERM domain containing 7
Gene Aliases
RGD1562057
Chromosome Location
Chr.X: 138097510 - 138149678 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008773574.1 XP_008771796.1 7 - 8 998 98

View Full Product Details