Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl13
Entrez Gene ID
313445
Gene Name
kelch-like family member 13
Gene Aliases
-
Chromosome Location
Chr.X: 121578965 - 121735007 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002730180.4 XP_002730226.1 2 - 3 619 73
XM_003752130.3 XP_003752178.1 3 - 4 2399 73
XM_006257443.2 XP_006257505.1 5 - 6 539 73
XM_006257444.2 XP_006257506.1 2 - 3 517 73
XM_006257445.2 XP_006257507.1 4 - 5 473 73
GenBank mRNA FQ228246.1 - 5 - 6 501 73

View Full Product Details