Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rhox5
Entrez Gene ID
24631
Gene Name
reproductive homeobox 5
Gene Aliases
Pem
Chromosome Location
Chr.X: 123900363 - 123916988 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006257472.2 XP_006257534.1 3 - 4 895 83
XM_006257473.2 XP_006257535.1 3 - 4 511 83
XM_006257475.1 XP_006257537.1 4 - 5 639 83
XM_008773400.1 XP_008771622.1 4 - 5 790 83
GenBank mRNA FQ228987.1 - 3 - 4 529 83
S82627.1 - 2 - 3 431 83

View Full Product Details