The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024897.1 | NP_001020068.1 | 4 - 5 | 996 | 139 |
GenBank mRNA | BC097385.1 | - | 4 - 5 | 996 | 139 |