The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031102.2 | NP_112364.1 | 5 - 6 | 500 | 93 |
XM_006229161.2 | XP_006229223.1 | 5 - 6 | 981 | 93 | |
GenBank mRNA | BC084727.1 | - | 4 - 5 | 492 | 93 |
CB577548.1 | - | 5 - 6 | 500 | 93 | |
DQ403026.1 | - | 4 - 5 | 416 | 93 | |
FQ211765.1 | - | 5 - 6 | 518 | 93 | |
FQ221110.1 | - | 5 - 6 | 518 | 93 | |
FQ222651.1 | - | 5 - 6 | 518 | 93 | |
FQ228615.1 | - | 5 - 6 | 521 | 93 | |
M20156.1 | - | 4 - 5 | 486 | 93 |