Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Selv
Entrez Gene ID
499113
Gene Name
selenoprotein V
Gene Aliases
RGD1562943
Chromosome Location
Chr.1: 85496940 - 85504189 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC158767.1 - 2 - 3 674 69

View Full Product Details