Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam98c
Entrez Gene ID
292764
Gene Name
family with sequence similarity 98, member C
Gene Aliases
RGD1303117
Chromosome Location
Chr.1: 88095240 - 88098816 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198584.1 NP_001185513.1 4 - 5 561 124
NR_037147.1 3 - 4 636 124
XM_006228717.2 XP_006228779.1 4 - 5 591 124
GenBank mRNA BC079080.1 - 3 - 4 599 124
CO564588.1 - 2 - 3 338 124
CO570848.1 - 4 - 5 561 124

View Full Product Details