Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd37
Entrez Gene ID
29185
Gene Name
CD37 molecule
Gene Aliases
-
Chromosome Location
Chr.1: 101230470 - 101236050 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017124.1 NP_058820.1 2 - 3 200 81
XM_006228981.2 XP_006229043.1 3 - 4 393 81
GenBank mRNA BC059144.1 - 2 - 3 241 81
FQ230385.1 - 2 - 3 218 81
X53517.1 - 2 - 3 200 81

View Full Product Details