Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Svip
Entrez Gene ID
499157
Gene Name
small VCP/p97-interacting protein
Gene Aliases
RGD1565432
Chromosome Location
Chr.1: 107368117 - 107373807 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271084.1 NP_001258013.1 2 - 3 225 125

View Full Product Details