Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cldnd2
Entrez Gene ID
499140
Gene Name
claudin domain containing 2
Gene Aliases
RGD1566413
Chromosome Location
Chr.1: 98487631 - 98490315 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109143.1 NP_001102613.1 3 - 4 424 82

View Full Product Details