Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mrap
Entrez Gene ID
288271
Gene Name
melanocortin 2 receptor accessory protein
Gene Aliases
RGD1310648
Chromosome Location
Chr.11: 30904733 - 30915225 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001135834.1 NP_001129306.1 1 - 2 183 62

View Full Product Details