Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fsd2
Entrez Gene ID
308779
Gene Name
fibronectin type III and SPRY domain containing 2
Gene Aliases
RGD1305167
Chromosome Location
Chr.1: 143364867 - 143398124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191627.1 NP_001178556.1 8 - 9 1538 139
XM_006229440.2 XP_006229502.1 9 - 10 1742 139
XM_008759537.1 XP_008757759.1 9 - 10 2959 139
XM_008759538.1 XP_008757760.1 10 - 11 2132 139

View Full Product Details