Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Homer2
Entrez Gene ID
29547
Gene Name
homer homolog 2 (Drosophila)
Gene Aliases
Vesl-2
Chromosome Location
Chr.1: 143443300 - 143535579 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB007690.1 - 4 - 5 480 71

View Full Product Details