Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mrpl46
Entrez Gene ID
293054
Gene Name
mitochondrial ribosomal protein L46
Gene Aliases
-
Chromosome Location
Chr.1: 140469557 - 140477634 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013068.1 NP_001013086.1 1 - 2 248 75
GenBank mRNA BC086407.1 - 1 - 2 248 75
FQ212933.1 - 1 - 2 244 75

View Full Product Details