Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr73
Entrez Gene ID
308751
Gene Name
WD repeat domain 73
Gene Aliases
Nmb
Chromosome Location
Chr.1: 142711792 - 142720292 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107525.2 NP_001100995.1 4 - 5 334 102
XM_006229430.2 XP_006229492.1 3 - 4 680 102
GenBank mRNA BC158658.1 - 4 - 5 334 102

View Full Product Details