Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tacc1
Entrez Gene ID
306562
Gene Name
transforming, acidic coiled-coil containing protein 1
Gene Aliases
Tacc1a
Chromosome Location
Chr.16: 71596970 - 71681126 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006253318.2 XP_006253380.1 10 - 11 1201 100
XM_008771354.1 XP_008769576.1 12 - 13 2197 100

View Full Product Details