Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Htra4
Entrez Gene ID
306564
Gene Name
HtrA serine peptidase 4
Gene Aliases
RGD1306242
Chromosome Location
Chr.16: 71784823 - 71802765 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107321.1 NP_001100791.1 6 - 7 1261 140
XM_008771355.1 XP_008769577.1 6 - 7 4425 140

View Full Product Details