The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017040.1 | NP_058736.1 | 5 - 6 | 1041 | 112 |
GenBank mRNA | BC085926.1 | - | 5 - 6 | 1025 | 112 |
FQ225122.1 | - | 1 - 2 | 77 | 112 | |
FQ231171.1 | - | 1 - 2 | 130 | 112 | |
M23591.1 | - | 5 - 6 | 1041 | 112 | |
X14087.1 | - | 5 - 6 | 874 | 112 | |
X16044.1 | - | 5 - 6 | 776 | 112 |