The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001008371.1 | NP_001008372.1 | 3 - 4 | 417 | 135 |
XM_006231042.1 | XP_006231104.1 | 3 - 4 | 514 | 135 | |
GenBank mRNA | BC086542.1 | - | 3 - 4 | 417 | 135 |