The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014106.1 | NP_001014128.1 | 3 - 4 | 596 | 169 |
GenBank mRNA | BC079125.1 | - | 3 - 4 | 596 | 169 |
FQ219206.1 | - | 3 - 4 | 564 | 169 |